Learn More
Azzota Corp hcl lamp Ytterbium 1.5in
Catalog No. 5000887175
Encompass_Preferred
1 5 Ytterbium (Yb) Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm
Catalog No. 50-008-87175 Supplier Azzota Corp Supplier No. SHC15YB
1 5 Ytterbium (Yb) Hollow Cathode Lamp Self-Reversal Diameter 37mm Length 160mm