Learn More
Aviva Systems Biology Corp HCLS1 Antibody - N-terminal region (ARP31928_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP31928_P050
This is a rabbit polyclonal antibody against HCLS1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human HCLS1. Alternate names: HS1, CTTNLApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54376