Learn More
ATCC HCM-BROD-0047-C71; Cancer Model Recurrent; Glioblastoma Brain; Human (Homo sapiens)
Catalog No. 5000549960
Encompass
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-BROD-0047-C71 (ATCC No. PDM-23) was isolated from recurrent glioblastoma of brain tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.
Catalog No. 50-005-49960 Supplier ATCC Supplier No. PDM23
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-BROD-0047-C71 (ATCC No. PDM-23) was isolated from recurrent glioblastoma of brain tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.