Learn More
ATCC HCM-BROD-0229-C25; Cancer Model Metastatic; Adenocarcinoma of pancreas; Human (Homo sapiens)
Catalog No. 5000550091
Encompass
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-BROD-0229-C25 (ATCC No. PDM-168) was isolated from metastatic adenocarcinoma of pancreas tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the tumor-derived organoid model is available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.
Catalog No. 50-005-50091 Supplier ATCC Supplier No. PDM168
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-BROD-0229-C25 (ATCC No. PDM-168) was isolated from metastatic adenocarcinoma of pancreas tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the tumor-derived organoid model is available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.