Learn More
ATCC HCM-CSHL-0063-C18; Cancer Model Primary; Adenocarcinoma Sigmoid Colon; Human (Homo sapiens)
Catalog No. 5000550242
Encompass
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0063-C18 (ATCC No. PDM-7) was isolated from primary adenocarcinoma of large intestine tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.
Catalog No. 50-005-50242 Supplier ATCC Supplier No. PDM7
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0063-C18 (ATCC No. PDM-7) was isolated from primary adenocarcinoma of large intestine tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.