Learn More
ATCC HCM-CSHL-0183-C25; Cancer Model Primary; Pancreatic cancer; Human (Homo sapiens)
Catalog No. 5000550398
Encompass
A patient-derived next-generation organoid cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0183-C25 (ATCC No. PDM-204) was isolated from a primary adenocarcinoma of the pancreas. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.
Catalog No. 50-005-50398 Supplier ATCC Supplier No. PDM204
A patient-derived next-generation organoid cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0183-C25 (ATCC No. PDM-204) was isolated from a primary adenocarcinoma of the pancreas. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.