Learn More
ATCC HCM-CSHL-0727-C18; Cancer Model; Primary; Colon cancer; Human (Homo sapiens)
Catalog No. 5000549746
Encompass
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0727-C18 (ATCC No. PDM-420) was isolated from primary colon cancer tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.
Catalog No. 50-005-49746 Supplier ATCC Supplier No. PDM420
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0727-C18 (ATCC No. PDM-420) was isolated from primary colon cancer tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.