Learn More
ATCC HCM-CSHL-0730-C25; Cancer Model; Primary; Pancreatic cancer; Human (Homo sapiens)
Catalog No. 5000549987
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0730-C25 (ATCC No. PDM-423) was isolated from primary pancreatic cancer tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the tumor-derived organoid model is available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.
Catalog No. 50-005-49987 Supplier ATCC Supplier No. PDM423
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0730-C25 (ATCC No. PDM-423) was isolated from primary pancreatic cancer tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the tumor-derived organoid model is available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.