Learn More
Lonza Walkersville HCM SingleQuots Kit
Catalog No. NC9700047
Encompass
The HCM SingleQuots Kit consists of individual vials containing the key labile ingredients required to combine with the HBM Basal Medium (Sold Seperately, CC-3199) in support of primary hepatocyte cultures. Once combinded together these reagents formulate a highly reproducible maintenance medium for primary hepatocytes from all species. The final medium is serum-free.Benefits:Optimized for Hepatocyte Cell HealthCompatible with all species HepatocytesContent:1 x HCM Singlequots Growth Supplement Pack (CC-4182)
Catalog No. NC9700047 Supplier Lonza Walkersville Supplier No. CC4182
The HCM SingleQuots Kit consists of individual vials containing the key labile ingredients required to combine with the HBM Basal Medium (Sold Seperately, CC-3199) in support of primary hepatocyte cultures. Once combinded together these reagents formulate a highly reproducible maintenance medium for primary hepatocytes from all species. The final medium is serum-free.Benefits:Optimized for Hepatocyte Cell HealthCompatible with all species HepatocytesContent:1 x HCM Singlequots Growth Supplement Pack (CC-4182)