Learn More
ATCC HCM-WCMC-0673-C56-B; Cancer model; Primary; Ovarian cancer; Human (Homo sapiens)
Catalog No. 5000550239
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
A patient-derived next-generation organoid cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-WCMC-0673-C56-B (ATCC No. PDM-514) was isolated from primary ovarian cancer tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.
Catalog No. 50-005-50239 Supplier ATCC Supplier No. PDM514
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
A patient-derived next-generation organoid cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-WCMC-0673-C56-B (ATCC No. PDM-514) was isolated from primary ovarian cancer tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.