Learn More
ATCC HCM-WCMC-0749-C34; Cancer model; Primary; Lung cancer; Human (Homo sapiens)
Catalog No. 5000550241
Encompass
A patient-derived next-generation organoid cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-WCMC-0749-C34 (ATCC No. PDM-683) was isolated from primary lung cancer tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Please check the GDC for the availability of any data for this model.
Catalog No. 50-005-50241 Supplier ATCC Supplier No. PDM683
A patient-derived next-generation organoid cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-WCMC-0749-C34 (ATCC No. PDM-683) was isolated from primary lung cancer tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Please check the GDC for the availability of any data for this model.