Learn More
ATCC HCM-BROD-0011-C71; Cancer Model Primary; Glioblastoma Brain; Human (Homo sapiens)
Catalog No. 502381888
Encompass
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-BROD-0011-C71 (ATCC No. PDM-18) was isolated from primary glioblastoma of brain tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.
Catalog No. 50-238-1888 Supplier ATCC Supplier No. PDM18
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-BROD-0011-C71 (ATCC No. PDM-18) was isolated from primary glioblastoma of brain tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.