Learn More
ATCC HCM-CSHL-0058-C34; Cancer Model Primary; Adenocarcinoma Right upper lobe lung; Human (Homo sapiens)
Catalog No. 502382508
Encompass
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0058-C34 (ATCC No. PDM-3) was isolated from primary adenocarcinoma of lung tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.
Catalog No. 50-238-2508 Supplier ATCC Supplier No. PDM3
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0058-C34 (ATCC No. PDM-3) was isolated from primary adenocarcinoma of lung tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.