Learn More
ATCC HCM-CSHL-0062-C18; Primary Adenocarcinoma Rectosigmoid junction; Human (Homo sapiens)
Catalog No. 502382664
Encompass
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0062-C18 (ATCC No. PDM-6) was isolated from primary adenocarcinoma of large intestine tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.
Catalog No. 50-238-2664 Supplier ATCC Supplier No. PDM6
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0062-C18 (ATCC No. PDM-6) was isolated from primary adenocarcinoma of large intestine tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.