Learn More
ATCC HCM-CSHL-0065-C20; Cancer Model Metastatic; Adenocarcinoma of rectum; Human (Homo sapiens)
Catalog No. 502383748
Encompass
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0065-C20 (ATCC No. PDM-9) was isolated from primary metastatic adenocarcinoma of large intestine tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.
Catalog No. 50-238-3748 Supplier ATCC Supplier No. PDM9
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0065-C20 (ATCC No. PDM-9) was isolated from primary metastatic adenocarcinoma of large intestine tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.