Learn More
ATCC HCM-CSHL-0089-C25; Cancer Model Primary; Pancreatic Ductal Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502381984
Encompass
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0089-C25 (ATCC No. PDM-36) was isolated from primary adenocarcinoma of pancreas tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.
Catalog No. 50-238-1984 Supplier ATCC Supplier No. PDM36
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0089-C25 (ATCC No. PDM-36) was isolated from primary adenocarcinoma of pancreas tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.