Learn More
Promocell GmbH PromoCell HCMEC-c 500,000 Human Cardiac Microvascular Endothelial Cells, cryopreserved
Catalog No. 50305994
Encompass
Primary Human Cardiac Microvascular Endothelial Cells (HCMEC) are isolated from heart ventricles from a single donor. The cells are routinely analyzed by immunofluorescent staining: they stain positive for CD31 and Willebrand factor and negative for smooth muscle alpha-actin.HCMEC play an important role in the physiological regulation of coronary blood flow and capillary exchange. Medical conditions such as hypertrophy, ischemia, hypertension, and diabetes mellitus are associated with endothelial dysfunction.
Catalog No. 50-305-994 Supplier Promocell GmbH Supplier No. C12285
Primary Human Cardiac Microvascular Endothelial Cells (HCMEC) are isolated from heart ventricles from a single donor. The cells are routinely analyzed by immunofluorescent staining: they stain positive for CD31 and Willebrand factor and negative for smooth muscle alpha-actin.HCMEC play an important role in the physiological regulation of coronary blood flow and capillary exchange. Medical conditions such as hypertrophy, ischemia, hypertension, and diabetes mellitus are associated with endothelial dysfunction.