Learn More
ACROBiosystems HCoV-229E Nucleocapsid protein, His Tag
Catalog No. 502055130
Encompass_Preferred
Recombinant Protein;1MG;HCoV-229E Nucleocapsid, His Tag (NUN-V5147) is expressed from E.coli cells. It contains AA Met 1 - Asn 389 (Accession # P15130-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 45.3 kDa.
Catalog No. 50-205-5130 Supplier ACROBiosystems Supplier No. NUNV51471MG
Recombinant Protein;1MG;HCoV-229E Nucleocapsid, His Tag (NUN-V5147) is expressed from E.coli cells. It contains AA Met 1 - Asn 389 (Accession # P15130-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 45.3 kDa.