Learn More
ACROBiosystems ACROBiosystems HCoV-229E Spike protein, His Tag
Catalog No. 502107092
Encompass_Preferred
Recombinant Protein;50UG;HCoV-229E Spike Protein, His Tag (SPN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 16 - Pro 1115 (Accession # P15423-1 (TI 871-872 PP)).The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag at the N-terminus. Proline substitutions TI 871-872 PP are introduced to stabilize the trimeric prefusion state of the spike protein.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 125.7 kDa. The protein migrates as 150-200 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7092 Supplier ACROBiosystems Supplier No. SPNH52H350UG
Recombinant Protein;50UG;HCoV-229E Spike Protein, His Tag (SPN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 16 - Pro 1115 (Accession # P15423-1 (TI 871-872 PP)).The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag at the N-terminus. Proline substitutions TI 871-872 PP are introduced to stabilize the trimeric prefusion state of the spike protein.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 125.7 kDa. The protein migrates as 150-200 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.