Learn More
Sino Biological Human coronavirus (HCoV-HKU1)Nucleocapsid Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40642T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human coronavirus (HCoV-HKU1) Nucleoprotein / NP ( Catalog#40642-V07E; YP_173242.1; Met1-Ala441). Coronavirus Nucleocapsid specific IgG was purified by Human coronavirus (HCoV-HKU1) Nucleoprotein / NP affinity chromatography.
Catalog No. 50-237-3975