Learn More
ACROBiosystems HCoV-OC43 Nucleocapsid protein, His Tag
Catalog No. 502055135
Encompass_Preferred
Recombinant Protein;100UG;HCoV-OC43 Nucleocapsid, His Tag (NUN-V5145) is expressed from E.coli cells. It contains AA Met 1 - Ile 448 (Accession # P33469-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 51 kDa.
Catalog No. 50-205-5135 Supplier ACROBiosystems Supplier No. NUNV5145100UG
Recombinant Protein;100UG;HCoV-OC43 Nucleocapsid, His Tag (NUN-V5145) is expressed from E.coli cells. It contains AA Met 1 - Ile 448 (Accession # P33469-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 51 kDa.