Learn More
ACROBiosystems HCoV-OC43 Nucleocapsid protein, His Tag
Supplier:  ACROBiosystems NUNV51451MG
Encompass_Preferred
Recombinant Protein;1MG;HCoV-OC43 Nucleocapsid, His Tag (NUN-V5145) is expressed from E.coli cells. It contains AA Met 1 - Ile 448 (Accession # P33469-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 51 kDa.
Catalog No. 50-205-5134