Learn More
Sino Biological Human coronavirus (HCoV-OC43) Spike S1+S2 Protein (ECD, His Tag), Biotinylated
Supplier:  Sino Biological 40607V08BB20
Encompass_Preferred
A DNA sequence encoding the human coronavirus (HCoV-OC43) spike protein (S1+S2 ECD) (AVR40344.1) (Met1-Pro1304) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. NC2072265