Learn More
Sino Biological Human coronavirus (HCoV-OC43) Spike Protein (S1+S2 ECD, His Tag)
Supplier:  Sino Biological 40607V08B100
Encompass_Preferred
A DNA sequence encoding the human coronavirus (HCoV-OC43) spike protein (S1+S2 ECD) (AVR40344.1) (Met1-Pro1304) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. NC1792225