Learn More
Enzo Life Sciences Coronavirus (hCoV-229E NP) antigen, (recombinant) (His-tag) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2890100
Encompass_Preferred
Purity: ≥95%. Formulation: Liquid. In 10 mM PBS, pH 7.2 with 3M urea. Source: Produced in E. coli. Recombinant protein fused to a His-tag. Composed of 359 amino acids containing immunodominant regions of coronavirus nucleoprotein. Long Term Storage: -20°C. Applications: ELISA .
Catalog No. 50-201-4100