Learn More
MilliporeSigma HCPRO DPTH FILTR NANOPOD NP6
Catalog No. NC3662129
Encompass
  • Fully synthetic filter minimizes extractables.
  • Hydrophilic wettability enhances performance.
  • Compact size with a diameter of 3.6 cm (1.4 in).
  • Low bacterial endotoxin levels at less than 0.25 EU/mL.
  • 9 filters per pack, with FSC certified packaging.
  • Operating temperature range of 4 to 40 °C.
  • Maximum operating pressure of 2.1 bar (30 psig).
  • Reduces feedstream volume requirements.
  • Designed for benchtop virus prefiltration and process development.
  • Post-use decontamination recommended for sterilization.
Catalog No. NC3662129 Supplier MilliporeSigma Supplier No. MX0SP06CL9
  • Fully synthetic filter minimizes extractables.
  • Hydrophilic wettability enhances performance.
  • Compact size with a diameter of 3.6 cm (1.4 in).
  • Low bacterial endotoxin levels at less than 0.25 EU/mL.
  • 9 filters per pack, with FSC certified packaging.
  • Operating temperature range of 4 to 40 °C.
  • Maximum operating pressure of 2.1 bar (30 psig).
  • Reduces feedstream volume requirements.
  • Designed for benchtop virus prefiltration and process development.
  • Post-use decontamination recommended for sterilization.