Learn More
Aviva Systems Biology Corp HCRTR1 Antibody - N-terminal region (ARP65327_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65327_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HCRTR1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human HCRTR1. Alternate names: OX1RApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rabbit, Rat
Catalog No. 50-288-474