Learn More
ABclonal Technology HCRTR1 Rabbit pAb
SDP
Catalog No. 5000104900
Encompass_Preferred
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein selectively binds the hypothalamic neuropeptide orexin A. A related gene (HCRTR2) encodes a G-protein coupled receptor that binds orexin A and orexin B.
Catalog No. 50-001-04900 Supplier ABclonal Technology Supplier No. A147401000UL
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein selectively binds the hypothalamic neuropeptide orexin A. A related gene (HCRTR2) encodes a G-protein coupled receptor that binds orexin A and orexin B.