Learn More
StatLab HCS AFB (+/-) Control Slides

Description
- Slides consist of formalin fixed, paraffin embedded tissue sections which contain known positive tissue elements.
- Use these slides with confidence to evaluate stain quality and performance in histology laboratories.
- Slides are created using non-adhesion or adhesion white glass slides with 90 degree corners.
- Each package includes a specific staining protocol.
Specifications
Specifications
| Content And Storage | Store at room temperature at 15°C to 30°C (59°F to 86°F) |
| Product Type | Control Slide |
| For Use With (Application) | HCS AFB |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.