Learn More
StatLab HCS Control Slides Fungus Adhesion Slides
Description
- HCS Control Slides consist of formalin fixed, paraffin embedded tissue sections which contain known positive tissue elements
- Use these slides with confidence to evaluate stain quality and performance in histology laboratories
- StatLab HCS control slides are created using adhesion white glass slides with 90° corners
- Each package includes a specific staining protocol
Specifications
Specifications
| Product Type | Control Slide |
| Quantity | 25/Pk. |