Learn More
StatLab HCS Control Slides Glycogen Adhesion Slides
SDP
Catalog No. 23640171
Change view
Click to view available options
Quantity:
25/Pk.
Catalog No. Quantity
23-640-171 25/Pk.
1 options
Catalog No. 23-640-171 Supplier StatLab Supplier No. CS021PS25
Add to Cart
Add to Cart
HCS Control Slides Glycogen Adhesion Slides
  • HCS Control Slides consist of formalin fixed, paraffin embedded tissue sections which contain known positive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • StatLab HCS control slides are created using adhesion white glass slides with 90° corners
  • Each package includes a specific staining protocol
Specifications
Product Type Control Slide
Quantity 25/Pk.