Learn More
StatLab HCS Mucin Control Slides

Description
- Slides consist of formalin fixed, paraffin embedded tissue sections which contain known positive tissue elements
- Use slides with confidence to evaluate stain quality and performance in histology laboratories
- HCS control slides are created using non-adhesion or adhesion white glass slides with 90 degree corners
- Tissue: Known positive
- Special Stain or Antibody: Mucicarmine Stain
- Each package includes a specific staining protocol
Specifications
Specifications
| Product Type | Control Slide |
| For Use With (Application) | HCS Mucin |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.