Learn More
Sigma Aldrich Fine Chemicals Biosciences Human Colorectal Carcinoma Cells (HCT 116) | Epithelial | Tumorigenic, Metastatic | 18 Hour Doubling Time | 1 vial
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 910910051VL
Encompass
HCT 116 colorectal carcinoma cells | Derived from: Adult human male | Cell type: Epithelial | Growth: Adherent monolayer | Doubling time: ~18 hours | Tumorigenic in mice: Yes | Metastatic in mice: Yes | Keywords: human, colorectal carcinoma, epithelial, adherent, tumorigenic, metastatic, doubling time
Catalog No. NC2281722