Learn More
Invivogen HCT116-Dual Cells, 3-7 x 10e6 cells
SDP
Catalog No. NC1044323
Encompass
HCT116-Dual cells are adherent epithelial cells that have been derived from the human HCT116 colorectal carcinoma cell line by stable integration of two inducible reporter constructs. HCT116-Dual™ cells express numerous pattern recognition receptors (PRRs), including the NOD-like receptors (NLRs) NOD1 and NOD2, the RIG-I-like receptor (RLR) RIG-I, and the Toll-like receptors (TLRs) TLR3 and TLR5 but not TLR2 or TLR4.
Catalog No. NC1044323 Supplier Invivogen Supplier No. HCTDNFIS
HCT116-Dual cells are adherent epithelial cells that have been derived from the human HCT116 colorectal carcinoma cell line by stable integration of two inducible reporter constructs. HCT116-Dual™ cells express numerous pattern recognition receptors (PRRs), including the NOD-like receptors (NLRs) NOD1 and NOD2, the RIG-I-like receptor (RLR) RIG-I, and the Toll-like receptors (TLRs) TLR3 and TLR5 but not TLR2 or TLR4.