Learn More
ATCC HCT116-SLC25A23-KO-c2; Colorectal carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL3488
Encompass
The HCT116-SLC25A23-KO-c2 cell line contains a -2 bp deletion in SLC25A23 gene and combined with the parental cell line and other HCT116 derivatives can be used in cancer drug screening.
Catalog No. 50-238-2731