Learn More
ATCC HCT116-SLC27A2-KO-c7; Colorectal carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL3509
Encompass
The HCT116-SLC27A2-KO-c7 cell line contains a -11 deletion/+1 bp insertion in SLC27A2 gene and combined with the parental cell line and other HCT116 derivatives can be used in cancer drug screening.
Catalog No. 50-238-1737
 Restricted Item