Learn More
ATCC HCT-15 Quantitative; purified DNA, 3 ug; Human
Catalog No. 502382278
Encompass
Purified genomic DNA isolated from the HCT-15 colon adenocarcinoma cell line (ATCC CCL-225). This product was quantified for oncology biomarker KRAS p.G13D via NGS and ddPCR.
Catalog No. 50-238-2278 Supplier ATCC Supplier No. CCL225DQ
Purified genomic DNA isolated from the HCT-15 colon adenocarcinoma cell line (ATCC CCL-225). This product was quantified for oncology biomarker KRAS p.G13D via NGS and ddPCR.