Learn More
Invivogen Soluble human CTLA-4 fused to an IgG1 Fc domain, 50ug
SDP
Catalog No. NC3782337
Encompass
Soluble human CTLA-4 (CD152) fused to an IgG1 Fc domainInvivoGen offers hCTLA4-Fc, a soluble human CTLA-4 (CD152) chimera protein generated by fusing the N-terminal extracellular domain of human CTLA-4 (aa 36-160) to the N-terminus of a human IgG1 Fc domain with a TEV (Tobacco Etch Virus) sequence linker.hCTLA-4-Fc has been produced in CHO cells and purified by affinity chromatography. It has an apparent molecular weight of ~47 kDa on an SDS‑PAGE gel.
Catalog No. NC3782337 Supplier Invivogen Supplier No. FCHCTLA4
Soluble human CTLA-4 (CD152) fused to an IgG1 Fc domainInvivoGen offers hCTLA4-Fc, a soluble human CTLA-4 (CD152) chimera protein generated by fusing the N-terminal extracellular domain of human CTLA-4 (aa 36-160) to the N-terminus of a human IgG1 Fc domain with a TEV (Tobacco Etch Virus) sequence linker.hCTLA-4-Fc has been produced in CHO cells and purified by affinity chromatography. It has an apparent molecular weight of ~47 kDa on an SDS‑PAGE gel.