Learn More
Apexbio Technology LLC HCTU 330645-87-9 25g
SDP
Catalog No. 5000093607
Encompass_Preferred
HCTU is a uronium-based peptide coupling reagent It is designed to facilitate peptide bond formation by reacting with the carboxyl group of amino acids or peptides to form an activated ester intermediate which then couples with amino groups to enable amide linkage formation HCTU exerts its activity primarily through activation of carboxyl groups promoting controlled amide bond construction Based on these properties HCTU holds research potential in peptide synthesis drug discovery protein engineering and preparation of biomaterials incorporating peptide motifs
Catalog No. 50-000-93607 Supplier Apexbio Technology LLC Supplier No. A702725000
HCTU is a uronium-based peptide coupling reagent It is designed to facilitate peptide bond formation by reacting with the carboxyl group of amino acids or peptides to form an activated ester intermediate which then couples with amino groups to enable amide linkage formation HCTU exerts its activity primarily through activation of carboxyl groups promoting controlled amide bond construction Based on these properties HCTU holds research potential in peptide synthesis drug discovery protein engineering and preparation of biomaterials incorporating peptide motifs