Learn More
Creative Biomart Recombinant HCV Core nucleocapsid Antigen, b-galactosidase-tagged, Rhodamine-Labled
Supplier:  Creative Biomart HCV157
Encompass_Preferred
The E. coli derived recombinant protein contains the HCV core nucleocapsid immunodominant regions, amino acids 2-192. The Rhodamine labeled protein is fused with b-galactosidase (114 kDa) at N-terminu
Catalog No. 50-208-4535