Learn More
Sino Biological HCV HCV-E2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 40280RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Hepatitis C virus (HCV-1a) E2 ( Catalog#40280-V08H; NP_751921.1; Glu1-Glu278). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-2416