Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HCV-IN-31 | 1998705-62-6 | 99.7% | 312.30 | 100 MG
SDP

Catalog No. 5000348119
Encompass_Preferred

HCV-IN-31 is an HCV inhibitor with an EC50/EC95 of 15.7 μM for HCV replicon. This compound is presented as a white to off-white solid with high purity, making it suitable for research applications related to infection, inflammation, and hepatitis C virus.

  • Inhibits HCV replicon activity
  • High purity of 99.69%
  • Provided as a solid, white to off-white compound
  • Optimal storage conditions for solid form: 4°C, protected from light
  • Optimal storage conditions for solvent solutions: -80°C for up to 6 months; -20°C for up to 1 month, protected from light
  • Soluble in DMSO

Catalog No. 50-003-48119 Supplier Medchemexpress LLC Supplier No. HY138305100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HCV-IN-31 is an HCV inhibitor with an EC50/EC95 of 15.7 μM for HCV replicon. This compound is presented as a white to off-white solid with high purity, making it suitable for research applications related to infection, inflammation, and hepatitis C virus.

  • Inhibits HCV replicon activity
  • High purity of 99.69%
  • Provided as a solid, white to off-white compound
  • Optimal storage conditions for solid form: 4°C, protected from light
  • Optimal storage conditions for solvent solutions: -80°C for up to 6 months; -20°C for up to 1 month, protected from light
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.