Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HCV-IN-31 | 1998705-62-6 | 99.7% | 312.30 | 25 MG
SDP

Catalog No. 5000352429
Encompass_Preferred

HCV-IN-31 (compound 4) is a potent inhibitor of the Hepatitis C Virus (HCV) replicon, demonstrating an EC50/EC95 of 15.7 μM. This compound is provided as a white to off-white solid, suitable for various research applications.

  • High purity of 99.7%
  • Molecular weight: 312.30
  • Molecular formula: C12H17FN6O3
  • CAS number: 1998705-62-6
  • Recommended storage: 4°C, protect from light
  • In solvent storage: -80°C for 6 months, -20°C for 1 month (protect from light)

Catalog No. 50-003-52429 Supplier Medchemexpress LLC Supplier No. 25MGHY138305
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HCV-IN-31 (compound 4) is a potent inhibitor of the Hepatitis C Virus (HCV) replicon, demonstrating an EC50/EC95 of 15.7 μM. This compound is provided as a white to off-white solid, suitable for various research applications.

  • High purity of 99.7%
  • Molecular weight: 312.30
  • Molecular formula: C12H17FN6O3
  • CAS number: 1998705-62-6
  • Recommended storage: 4°C, protect from light
  • In solvent storage: -80°C for 6 months, -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.