Learn More
Medchemexpress LLC Hcv-in-31 | 1998705-62-6 | 99.7% | 312.30 g/mol | C12H17FN6O3 | 5 MG
SDP
Catalog No. 5000223410
Encompass_Preferred
HCV-IN-31 is a small-molecule inhibitor of hepatitis C virus (HCV) replication (compound 4). Reported activity in an HCV replicon shows an EC50/EC95 of 15.7 μM. The compound is supplied as a solid for research use only; molecular formula C12H17FN6O3, molecular weight 312.30, and high reported purity.
  • Inhibits HCV replicon with EC50/EC95 of 15.7 μM.
  • High purity (approximately 99.7%).
  • White to off-white solid suitable for analytical and biological studies.
  • Storage conditions: 4°C; in solvent store -80°C (6 months) or -20°C (1 month).
  • Available as small solid quantities and as a 10 mM solution in DMSO for assays.
Catalog No. 50-002-23410 Supplier Medchemexpress LLC Supplier No. HY1383055MG
HCV-IN-31 is a small-molecule inhibitor of hepatitis C virus (HCV) replication (compound 4). Reported activity in an HCV replicon shows an EC50/EC95 of 15.7 μM. The compound is supplied as a solid for research use only; molecular formula C12H17FN6O3, molecular weight 312.30, and high reported purity.
  • Inhibits HCV replicon with EC50/EC95 of 15.7 μM.
  • High purity (approximately 99.7%).
  • White to off-white solid suitable for analytical and biological studies.
  • Storage conditions: 4°C; in solvent store -80°C (6 months) or -20°C (1 month).
  • Available as small solid quantities and as a 10 mM solution in DMSO for assays.