Learn More
Medchemexpress LLC Hcv-in-31 | 1998705-62-6 | 99.7% | C12H17FN6O3 | 10MG
SDP
Catalog No. 5000260888
Encompass_Preferred
HCV-IN-31 is a small-molecule inhibitor of hepatitis C virus (HCV) replication supplied for laboratory research use. It shows activity in HCV replicon assays and is provided as a high-purity solid for in vitro biochemical and cell-based studies.
  • Reported EC50/EC95 of 15.7 μM against HCV replicon.
  • High reported purity (≈99.7%) suitable for analytical and biological testing.
  • Solid, white to off-white appearance for straightforward handling and weighing.
  • Documentation available including data sheet, certificate of analysis, and safety data sheet.
  • Storage guidance provided for solid and solvent conditions to preserve stability.
Catalog No. 50-002-60888 Supplier Medchemexpress LLC Supplier No. HY13830510MG
HCV-IN-31 is a small-molecule inhibitor of hepatitis C virus (HCV) replication supplied for laboratory research use. It shows activity in HCV replicon assays and is provided as a high-purity solid for in vitro biochemical and cell-based studies.
  • Reported EC50/EC95 of 15.7 μM against HCV replicon.
  • High reported purity (≈99.7%) suitable for analytical and biological testing.
  • Solid, white to off-white appearance for straightforward handling and weighing.
  • Documentation available including data sheet, certificate of analysis, and safety data sheet.
  • Storage guidance provided for solid and solvent conditions to preserve stability.