Learn More
Medchemexpress LLC HCV-IN-47 25g
SDP
Catalog No. 5000205039
Encompass_Preferred
HCV-IN-47 (Compound (R)-1) shows anti-hepatitis C virus (HCV) activity with an EC50 value of 0 032 M HCV-IN-47 is promising for research of hepatitis C virus pathogenesis and host-virus interaction[1]
Catalog No. 50-002-05039 Supplier Medchemexpress LLC Supplier No. 25GHYW103964
HCV-IN-47 (Compound (R)-1) shows anti-hepatitis C virus (HCV) activity with an EC50 value of 0 032 M HCV-IN-47 is promising for research of hepatitis C virus pathogenesis and host-virus interaction[1]