Learn More
Creative Biomart Recombinant HCV Antigen
Supplier:  Creative Biomart HCV176
Encompass_Preferred
Recombinant HCV antigen for lateral flow assay contains 2 HCV proteins. The mosaic HCV protein is composed of HCV core-NS3-NS4-NS5 which is used for colloid gold conjugate, and a protein cocktail t
Catalog No. 50-208-4554
 Restricted Item