Learn More
Creative Biomart Recombinant HCV Genotype 1b NS3 Antigen, b-galactosidase-tagged
Supplier:  Creative Biomart HCV179
Encompass_Preferred
The E. coli derived 22 kDa recombinant protein contains the HCV NS3 genotype 1b immunodominant regions, amino acids 1450-1643. The protein is fused with b-galactosidase (114 kDa) at N-terminus, pI 5.4
Catalog No. 50-208-4557