Learn More
Creative Biomart Recombinant HCV NS3 Antigen, His-tagged
Supplier:  Creative Biomart HCV185
Encompass_Preferred
Recombinant HCV NS3 Genotype 1a produced in E. coli is a non-glycosylated polypeptide chain having a molecular mass of 47.8 kDa and fused to a His tag at N-terminus.
Catalog No. 50-208-4563