Learn More
Creative Biomart Recombinant HCV NS3 Antigen, His-tagged, Biotin-Labeled
Supplier:  Creative Biomart HCV199
Encompass_Preferred
The E. coli derived recombinant Biotin Labeled protein contains the HCV NS3 immunodominant regions, a.a. 1450-1643, 22 kDa. The Biotin labeled protein is fused with a 6 His-Tag at N-terminus.
Catalog No. 50-208-4577